2-Amino-3-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A740006-1G A740006-5G
77158-778EA 86.52 USD
77158-778 77158-780
2-Amino-3-nitrobenzonitrile 98%
2-Amino-3-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MW: 163.13 g/mol
MDL Number: MFCD18389917
CAS Number: 87331-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR