2-Fluoro-4-methoxybenzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Fluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-4-methoxyphenylboronic acid pinacol ester

A739414-1G A739414-250MG A739414-5G
77158-772EA 43.26 USD
77158-772 77158-774 77158-776
2-Fluoro-4-methoxybenzeneboronic acid pinacol ester 98%
2-Fluoro-4-methoxybenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₃
MW: 252.09 g/mol
MDL Number: MFCD16996299
CAS Number: 628692-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR