5,5-Dimethyl-2-(naphthalen-2-yl)-1,3,2-dioxaborinane 97%
Supplier: Ambeed
|
Warning
|
A737584-1G
A737584-25G
A737584-5G
77158-758EA
24
USD
77158-758
77158-760
77158-762
5,5-Dimethyl-2-(naphthalen-2-yl)-1,3,2-dioxaborinane 97%
5,5-Dimethyl-2-(naphthalen-2-yl)-1,3,2-dioxaborinane
Precaución: for Research Use Only
Formula:
C₁₅H₁₇BO₂ |
MDL Number:
MFCD28122664 CAS Number: 627906-96-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...