(1H-Benzoimidazol-2-ylmethyl)methylamine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(1H-Benzo[d]imidazol-2-yl)-N-methylmethanamine

A736566-100MG A736566-1G A736566-250MG
77158-752EA 44.29 USD
77158-752 77158-754 77158-756
(1H-Benzoimidazol-2-ylmethyl)methylamine 98%
(1H-Benzoimidazol-2-ylmethyl)methylamine

Precaución: for Research Use Only

Formula: C₉H₁₁N₃
MW: 161.21 g/mol
MDL Number: MFCD06738854
CAS Number: 98997-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR