Ethyl 3-amino-6-methylpicolinate 95%

Supplier: Ambeed
Warning

A736052-100MG A736052-1G A736052-250MG
77158-740EA 52.53 USD
77158-740 77158-742 77158-744
Ethyl 3-amino-6-methylpicolinate 95%
Ethyl 3-amino-6-methylpicolinate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MW: 180.20 g/mol
MDL Number: MFCD18382757
CAS Number: 908832-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR