(3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol 98%

Supplier: Ambeed
Warning

A735525-100MG A735525-1G A735525-250MG A735525-5G
77158-732EA 36.05 USD
77158-732 77158-734 77158-736 77158-738
(3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol 98%
(3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₈BClO₃
MW: 268.54 g/mol
MDL Number: MFCD18434467
CAS Number: 1485110-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR