1-[3-(Dimethylamino)propyl]-3-ethylurea 98%

Supplier: Ambeed
Warning

A734200-1G A734200-250MG A734200-5G
77158-728EA 61.8 USD
77158-728 77158-730 77614-258
1-[3-(Dimethylamino)propyl]-3-ethylurea 98%
1-[3-(Dimethylamino)propyl]-3-ethylurea

Precaución: for Research Use Only

Formula: C₈H₁₉N₃O
MDL Number: MFCD00027752
CAS Number: 32897-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR