Di-tert-butyl 2-(aminomethyl)piperazine-1,4-dicarboxylate 95%

Supplier: Ambeed
Warning

A731669-100MG A731669-250MG A731669-1G
77158-700EA 133.9 USD
77158-700 77158-702 77614-244
Di-tert-butyl 2-(aminomethyl)piperazine-1,4-dicarboxylate 95%
Di-tert-butyl 2-(aminomethyl)piperazine-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₉N₃O₄
MW: 315.40 g/mol
MDL Number: MFCD24471267
CAS Number: 1256815-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR