Reparixin 98%

Supplier: Ambeed
Warning

A730164-10MG A730164-1MG A730164-25MG A730164-50MG A730164-5MG
77158-690EA 51.01 USD
77158-690 77158-692 77158-694 77158-696 77158-698
Reparixin 98%
Reparixin

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₃S
MDL Number: MFCD18633292
CAS Number: 266359-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR