5-Bromo-3-(difluoromethyl)-2-methoxypyridine 95%

Supplier: Ambeed
Warning

A725437-1G A725437-250MG A725437-50MG
77158-652EA 358.44 USD
77158-652 77158-654 77686-724
5-Bromo-3-(difluoromethyl)-2-methoxypyridine 95%
5-Bromo-3-(difluoromethyl)-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₇H₆BrF₂NO
MW: 238.02 g/mol
MDL Number: MFCD23163928
CAS Number: 1254123-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR