Ethyl 2-amino-3-(4-hydroxyphenyl)-2-methylpropanoate 95%

Supplier: Ambeed
Warning

A725169-100MG A725169-250MG
77158-644EA 18.54 USD
77158-644 77158-646
Ethyl 2-amino-3-(4-hydroxyphenyl)-2-methylpropanoate 95%
Ethyl 2-amino-3-(4-hydroxyphenyl)-2-methylpropanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₃
MW: 223.26 g/mol
MDL Number: MFCD10566154
CAS Number: 16323-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR