cis-Methyl 3-hydroxy-1-methyl-cyclobutanecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: cis-Methyl 3-hydroxy-1-methylcyclobutanecarboxylate

A724822-1G A724822-250MG
77158-640EA 501.61 USD
77158-640 77158-642
cis-Methyl 3-hydroxy-1-methyl-cyclobutanecarboxylate 98%
cis-Methyl 3-hydroxy-1-methyl-cyclobutanecarboxylate

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.168 g/mol
MDL Number: MFCD23105987
CAS Number: 626238-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR