2-Ethoxypyrimidin-4-ylamine 95+%

Supplier: Ambeed
Warning

A724198-1G A724198-250MG A724198-5G
77158-624EA 64 USD
77158-624 77158-626 77158-628
2-Ethoxypyrimidin-4-ylamine 95+%
2-Ethoxypyrimidin-4-ylamine

Precaución: for Research Use Only

Formula: C₆H₉N₃O
MDL Number: MFCD00234232
CAS Number: 3289-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR