tert-Butyl-4-(1H-indol-1-yl)piperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(1H-indol-1-yl)piperidine-1-carboxylate

A723904-250MG A723904-5G
77158-614EA 36.05 USD
77158-614 77158-616
tert-Butyl-4-(1H-indol-1-yl)piperidine-1-carboxylate 95%
tert-Butyl-4-(1H-indol-1-yl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₄N₂O₂
MW: 300.4 g/mol
MDL Number: MFCD09833914
CAS Number: 170364-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR