7-Aminoheptanoic acid hydrochloride 98%

Supplier: Ambeed
Warning

A719974-100G A719974-25G A719974-5G
77158-600EA 143.17 USD
77158-600 77158-602 77158-604
7-Aminoheptanoic acid hydrochloride 98%
7-Aminoheptanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClNO₂
MW: 181.66 g/mol
MDL Number: MFCD12405602
CAS Number: 62643-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR