6-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole 97%

Supplier: Ambeed
Warning

A719740-1G A719740-250MG A719740-5G
77158-594EA 50.47 USD
77158-594 77158-596 77158-598
6-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole 97%
6-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂O
MW: 281.14 g/mol
MDL Number: MFCD22380213
CAS Number: 1158680-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR