(R)-tert-Butyl 5-methyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide 95%

Supplier: Ambeed
Warning

A718690-250MG A718690-5G A718690-1G
77158-588EA 46.35 USD
77158-588 77158-590 77614-210
(R)-tert-Butyl 5-methyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide 95%
(R)-tert-Butyl 5-methyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide

Precaución: for Research Use Only

Formula: C₈H₁₅NO₅S
MW: 237.27 g/mol
MDL Number: MFCD27999446
CAS Number: 863453-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR