Simeprevir ≥98%

Supplier: Ambeed
Warning

Synonyms: TMC 435350, TMC 435, (2R,3aR,11aS,12aR,14aR,Z)-N-(Cyclopropylsulphonyl)-2-((2-(4-isopropylthiazol-2-yl)-7-methoxy-8-methylquinolin-4-yl)oxy)-5-methyl-4,14-dioxo-2,3,3a,4,5,6,7,8,9,11a,12,13,14,14a-tetradecahydrocyclopenta[c]cyclopropa[g][1,6]diazacyclotetradecine-12a(1H)-carboxamide, (2R,3aR,10Z,11aS,12aR,14aR)-N-(Cyclopropylsulphonyl)-2,3,3a,4,5,6,7,8,9,11a,12,13,14,14a-tetradecahydro-2-[[7-methoxy-8-methyl-2-[4-(1-methylethyl)-2-thiazolyl]-4-quinolinyl]oxy]-5-methyl-4,14-dioxo-cyclopenta[c]cyclopropa[g][1,6]diazacyclotetradecine-12a(1H)-carboxamide

A713692-10MG A713692-1MG A713692-5MG
77158-576EA 47.38 USD
77158-576 77158-578 77158-580
Simeprevir ≥98%
Simeprevir

Precaución: for Research Use Only

Formula: C₃₈H₄₇N₅O₇S₂
MW: 749.96 g/mol
MDL Number: MFCD25563225
CAS Number: 923604-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR