1,3-Bis(4-fluorophenyl)urea 98%

Supplier: Ambeed
Warning

A713547-1G A713547-250MG A713547-5G
77158-570EA 42.23 USD
77158-570 77158-572 77158-574
1,3-Bis(4-fluorophenyl)urea 98%
1,3-Bis(4-fluorophenyl)urea

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂N₂O
MW: 248.23 g/mol
MDL Number: MFCD00017925
CAS Number: 370-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR