3-Chloro-2-hydroxypropyl acetate 95%

Supplier: Ambeed
Warning

A712867-100MG A712867-1G A712867-250MG A712867-5G
77158-544EA 22.66 USD
77158-544 77158-546 77158-548 77158-550
3-Chloro-2-hydroxypropyl acetate 95%
3-Chloro-2-hydroxypropyl acetate

Precaución: for Research Use Only

Formula: C₅H₉ClO₃
MW: 152.576 g/mol
MDL Number: MFCD01698128
CAS Number: 24573-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR