3-(6-Oxo-1,6-dihydro-3-pyridazinyl)benzonitrile 95%

Supplier: Ambeed
Danger

A710785-1G A710785-25G A710785-5G
77158-538EA 21 USD
77158-538 77158-540 77158-542
3-(6-Oxo-1,6-dihydro-3-pyridazinyl)benzonitrile 95%
3-(6-Oxo-1,6-dihydro-3-pyridazinyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₁H₇N₃O
MDL Number: MFCD16482247
CAS Number: 52240-08-1
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR