1,2-Dithiolane-4-carboxylic acid 98%

Supplier: Ambeed
Danger

Synonyms: Asparagusic acid

A710707-100MG A710707-10MG A710707-1MG A710707-250MG A710707-5MG A710707-50MG
77158-528EA 231.75 USD
77158-528 77158-530 77158-532 77158-534 77158-536 77684-627
1,2-Dithiolane-4-carboxylic acid 98%
1,2-Dithiolane-4-carboxylic acid

Precaución: For research use only.

Formula: C₄H₆O₂S₂
MW: 150.21 g/mol
MDL Number: MFCD01729684
CAS Number: 2224-02-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR