6-Bromo-2-methylpyrazolo[1,5-a]pyrimidine 98+%

Supplier: Ambeed
Warning

A709579-1G A709579-25G A709579-5G
77158-510EA 55.62 USD
77158-510 77158-512 77158-514
6-Bromo-2-methylpyrazolo[1,5-a]pyrimidine 98+%
6-Bromo-2-methylpyrazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
MDL Number: MFCD09863428
CAS Number: 916256-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR