1-Bromo-3-hydroxynaphthalene 95%

Supplier: Ambeed
Warning

A709452-100MG A709452-10G A709452-1G A709452-250MG A709452-25G A709452-5G A709452-100G
77158-498EA 6.18 USD
77158-498 77158-500 77158-502 77158-504 77158-506 77158-508 77573-302
1-Bromo-3-hydroxynaphthalene 95%
1-Bromo-3-hydroxynaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₇BrO
MW: 223.066 g/mol
MDL Number: MFCD01712260
CAS Number: 5498-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR