4-(Phenylethynyl)benzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(4-phenylethynyl-phenyl)-[1,3,2]dioxaborolane

A706216-100MG A706216-1G A706216-250MG A706216-5G
77158-486EA 26 USD
77158-486 77158-488 77158-490 77158-492
4-(Phenylethynyl)benzeneboronic acid pinacol ester 98%
4-(Phenylethynyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₂₁BO₂
MDL Number: MFCD16294498
CAS Number: 1190376-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR