2-Bromo-3-iodotoluene 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1-iodo-3-methylbenzene

A706032-100MG A706032-1G A706032-250MG A706032-25G A706032-5G
77158-478EA 10.3 USD
77158-478 77158-480 77158-482 77158-484 77552-152
2-Bromo-3-iodotoluene 95%
2-Bromo-3-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrI
MW: 296.9310 g/mol
MDL Number: MFCD11977389
CAS Number: 888214-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR