4-Chloro-2-(trifluoromethoxy)benzaldehyde 95%

Supplier: Ambeed
Warning

A702671-100MG A702671-1G A702671-250MG
77158-446EA 16.48 USD
77158-446 77158-448 77158-450
4-Chloro-2-(trifluoromethoxy)benzaldehyde 95%
4-Chloro-2-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MW: 224.56 g/mol
MDL Number: MFCD18393772
CAS Number: 1261442-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR