6-Cyano-2-naphthyl trifluoromethanesulfonate 95%

Supplier: Ambeed
Warning

A702237-1G A702237-250MG A702237-5G
77158-440EA 67 USD
77158-440 77158-442 77158-444
6-Cyano-2-naphthyl trifluoromethanesulfonate 95%
6-Cyano-2-naphthyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₂H₆F₃NO₃S
MDL Number: MFCD11505953
CAS Number: 145369-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR