2,6-Bis[3-(9H-carbazol-9-yl)phenyl]pyridine ≥99% (by HPLC), sublimated

Supplier: Ambeed
Warning

Synonyms: DCzPPy

A702119-100MG A702119-1G A702119-250MG
77158-434EA 67.98 USD
77158-434 77158-436 77158-438
2,6-Bis[3-(9H-carbazol-9-yl)phenyl]pyridine ≥99% (by HPLC), sublimated
2,6-Bis[3-(9H-carbazol-9-yl)phenyl]pyridine

Precaución: for Research Use Only

Formula: C₄₁H₂₇N₃
MW: 561.69 g/mol
MDL Number: MFCD20275106
CAS Number: 1013405-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR