3-Methylsulfonylpropylamine Hydrochloride 98%

Supplier: Ambeed
Warning

A694253-100MG A694253-250MG A694253-5G A694253-1G
77158-396EA 22 USD
77158-396 77158-398 77158-400 77686-677
3-Methylsulfonylpropylamine Hydrochloride 98%
3-Methylsulfonylpropylamine Hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₂ClNO₂S
MDL Number: MFCD09744144
CAS Number: 157825-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR