5-Bromo-3-chloro-2-fluoronitrobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1-chloro-2-fluoro-3-nitrobenzene

A694109-100MG A694109-10G A694109-1G A694109-250MG A694109-25G A694109-5G
77158-384EA 13 USD
77158-384 77158-386 77158-388 77158-390 77158-392 77901-609
5-Bromo-3-chloro-2-fluoronitrobenzene 97%
5-Bromo-3-chloro-2-fluoronitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFNO₂
MDL Number: MFCD24539370
CAS Number: 1435806-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR