tert-Butyl 3-azabicyclo[3.1.1]heptan-6-ylcarbamate 97%

Supplier: Ambeed
Warning

A693567-250MG A693567-100MG
77158-366EA 439 USD
77158-366 77893-388
tert-Butyl 3-azabicyclo[3.1.1]heptan-6-ylcarbamate 97%
tert-Butyl 3-azabicyclo[3.1.1]heptan-6-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD17168423
CAS Number: 1363382-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR