tert-Butyl 4-(methylsulfonyl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A692983-1G A692983-25G A692983-5G
77158-360EA 19.57 USD
77158-360 77158-362 77158-364
tert-Butyl 4-(methylsulfonyl)piperazine-1-carboxylate 95%
tert-Butyl 4-(methylsulfonyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₄S
MW: 264.34 g/mol
MDL Number: MFCD03942726
CAS Number: 164331-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR