3-Bromo-1-ethanesulfanylbenzene 98%

Supplier: Ambeed
Warning

A692947-1G A692947-250MG A692947-25G A692947-5G
77158-352EA 18 USD
77158-352 77158-354 77158-356 77158-358
3-Bromo-1-ethanesulfanylbenzene 98%
3-Bromo-1-ethanesulfanylbenzene

Precaución: for Research Use Only

Formula: C₈H₉BrS
MDL Number: MFCD10699173
CAS Number: 18184-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR