CLP-290 95%

Supplier: Ambeed
Warning

Synonyms: (Z)-5-Fluoro-2-((4-oxo-2-(tetrahydropyridazin-1(2H)-yl)thiazol-5(4H)-ylidene)methyl)phenyl pyrrolidine-1-carboxylate, [5-Fluoro-2-[(Z)-(2-hexahydropyridazin-1-yl-4-oxo-thiazol-5-ylidene)methyl]phenyl] pyrrolidine-1-carboxylate

A692338-10MG A692338-1MG A692338-25MG A692338-5MG
77158-344EA 55 USD
77158-344 77158-346 77158-348 77158-350
CLP-290 95%
CLP-290

Precaución: for Research Use Only

Formula: C₁₉H₂₁FN₄O₃S
MW: 404.46 g/mol
MDL Number: MFCD28166314
CAS Number: 1181083-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR