2-Bromo-3,5-dimethylphenol 98%

Supplier: Ambeed
Warning

A691185-1G A691185-250MG A691185-5G
77158-338EA 44.29 USD
77158-338 77158-340 77158-342
2-Bromo-3,5-dimethylphenol 98%
2-Bromo-3,5-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
MDL Number: MFCD22058629
CAS Number: 125237-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR