Ki20227 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-((6,7-Dimethoxyquinolin-4-yl)oxy)-2-methoxyphenyl)-3-(1-(thiazol-2-yl)ethyl)urea

A691021-10MG A691021-1MG A691021-25MG A691021-2MG A691021-5MG
77158-324EA 369.77 USD
77158-324 77158-326 77158-328 77158-330 77158-332
Ki20227 98%
Ki20227

Precaución: for Research Use Only

Formula: C₂₄H₂₄N₄O₅S
MW: 480.5 g/mol
MDL Number: MFCD12024693
CAS Number: 623142-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR