Ethyl-4-chloro-8-fluoroquinoline-3-carboxylate 98%

Supplier: Ambeed
Warning

A688729-1G A688729-5G
77158-310EA 77 USD
77158-310 77158-312
Ethyl-4-chloro-8-fluoroquinoline-3-carboxylate 98%

Precaución: for Research Use Only

Formula: C₁₂H₉ClFNO₂
MDL Number: MFCD00173364
CAS Number: 56824-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR