2,6-Di-O-methyl-β-cyclodextrine 98%

Supplier: Ambeed
Warning

Synonyms: Dimethyl-β-cyclodextrin, methylated β-cyclodextrins, Heptakis(2,6-di-o-methyl)-β-cyclodextrin, 2,6-Di-O-methyl-β-cyclodextrin

A688339-100G A688339-10G A688339-1G A688339-25G
77158-302EA 328.57 USD
77158-302 77158-304 77158-306 77158-308
2,6-Di-O-methyl-β-cyclodextrine 98%
2,6-Di-O-methyl-β-cyclodextrine

Precaución: for Research Use Only

Formula: C₅₆H₉₈O₃₅
MW: 1331.3563 g/mol
MDL Number: MFCD00011616
CAS Number: 51166-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR