2-Methyl-5-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1,3,4-oxadiazole 95+%
Supplier: Ambeed
|
Warning
|
A688257-1G
A688257-250MG
A688257-5G
77158-296EA
121.54
USD
77158-296
77158-298
77158-300
2-Methyl-5-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1,3,4-oxadiazole 95+%
2-Methyl-5-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1,3,4-oxadiazole
Precaución: for Research Use Only
Formula:
C₁₅H₁₉BN₂O₃ MW: 286.13 g/mol |
MDL Number:
MFCD18384931 CAS Number: 1056456-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...