trans-4-(2-Thienyl)-3-buten-2-one 95+%

Supplier: Ambeed
Warning

A686664-1G A686664-25G A686664-5G
77158-290EA 15.45 USD
77158-290 77158-292 77158-294
trans-4-(2-Thienyl)-3-buten-2-one 95+%
trans-4-(2-Thienyl)-3-buten-2-one

Precaución: for Research Use Only

Formula: C₈H₈OS
MW: 152.21 g/mol
MDL Number: MFCD00053051
CAS Number: 33603-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR