5-Chloro-2,4-difluorobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 5-Chloro-2,4-difluorobenzenesulphonylchloride

A685021-10G A685021-1G A685021-25G A685021-5G
77158-282EA 20.6 USD
77158-282 77158-284 77158-286 77158-288
5-Chloro-2,4-difluorobenzenesulfonyl chloride 98%
5-Chloro-2,4-difluorobenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₂Cl₂F₂O₂S
MW: 247.05 g/mol
MDL Number: MFCD01940426
CAS Number: 13656-57-0
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR