Ethyl 5-methyl-1,3,4-thiadiazole-2-carboxylate 98%

Supplier: Ambeed
Warning

A684498-100MG A684498-1G A684498-250MG A684498-5G
77158-274EA 31.93 USD
77158-274 77158-276 77158-278 77158-280
Ethyl 5-methyl-1,3,4-thiadiazole-2-carboxylate 98%
Ethyl 5-methyl-1,3,4-thiadiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂S
MW: 172.21 g/mol
MDL Number: MFCD03840446
CAS Number: 869108-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR