Darapladib 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-(Diethylamino)ethyl)-2-(2-((4-fluorobenzyl)thio)-4-oxo-4,5,6,7-tetrahydro-1H-cyclopenta[d]pyrimidin-1-yl)-N-((4'-(trifluoromethyl)-[1,1'-biphenyl]-4-yl)methyl)acetamide, N-[2-(diethylamino)ethyl]-2-[[(4-fluorophenyl)methyl]thio]-4,5,6,7-tetrahydro-4-oxo-N-[[4'-(trifluoromethyl)[1,1'-biphenyl]-4-yl]methyl]-

A684359-10MG A684359-1MG A684359-50MG A684359-5MG
77158-266EA 46.35 USD
77158-266 77158-268 77158-270 77158-272
Darapladib 98%
Darapladib

Precaución: for Research Use Only

Formula: C₃₆H₃₈F₄N₄O₂S
MW: 666.78 g/mol
MDL Number: MFCD18251452
CAS Number: 356057-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR