Dimethyl 4-nitroisophthalate 96%

Supplier: Ambeed
Warning

A684068-100MG A684068-1G
77158-262EA 39.14 USD
77158-262 77158-264
Dimethyl 4-nitroisophthalate 96%
Dimethyl 4-nitroisophthalate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₆
MW: 239.18 g/mol
MDL Number: MFCD00177948
CAS Number: 69048-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR