2,4-Difluorobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2,4-Difluorobenzene-1-sulphonyl chloride

A684027-100G A684027-10G A684027-1G A684027-25G A684027-5G
77158-252EA 73.13 USD
77158-252 77158-254 77158-256 77158-258 77158-260
2,4-Difluorobenzenesulfonyl chloride 98%
2,4-Difluorobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
MDL Number: MFCD00042481
CAS Number: 13918-92-8
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR