2,2,2-Trichloroethanol 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,2,2-Trichloroethan-1-ol
A683276-100G
A683276-25G
A683276-500G
77158-246EA
24.72
USD
77158-246
77158-248
77158-250
2,2,2-Trichloroethanol 98%
2,2,2-Trichloroethanol
Precaución: for Research Use Only
Formula:
C₂H₃Cl₃O MW: 149.4 g/mol |
MDL Number:
MFCD00004677 CAS Number: 115-20-8 UN: 3082 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...