3-Aminothietane 1,1-dioxide 95%

Supplier: Ambeed
Warning

A682874-100MG A682874-250MG A682874-1G
77158-244EA 135.96 USD
77158-244 77614-110 77686-654
3-Aminothietane 1,1-dioxide 95%
3-Aminothietane 1,1-dioxide

Precaución: for Research Use Only

Formula: C₃H₇NO₂S
MW: 121.16 g/mol
MDL Number: MFCD18445741
CAS Number: 88511-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR