3-Bromo-5-chloro-2-fluoroaniline 97%
Supplier: Ambeed
|
Warning
|
A681989-1G
A681989-250MG
A681989-25G
A681989-5G
77158-236EA
28
USD
77158-236
77158-238
77158-240
77158-242
3-Bromo-5-chloro-2-fluoroaniline 97%
3-Bromo-5-chloro-2-fluoroaniline
Precaución: for Research Use Only
Formula:
C₆H₄BrClFN |
MDL Number:
MFCD19382423 CAS Number: 1269232-95-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...