Methyl 5-aminopyrazine-2-carboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A681905-1G A681905-250MG A681905-5G
77158-230EA 46.35 USD
77158-230 77158-232 77158-234
Methyl 5-aminopyrazine-2-carboxylate hydrochloride 98%
Methyl 5-aminopyrazine-2-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₈ClN₃O₂
MW: 189.59 g/mol
MDL Number: MFCD18425624
CAS Number: 1417794-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR